Product Details

SNP ID
rs148949332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:7831935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAAGTCAGCCTGCACATGGTTG[G/T]CACGGCCAAAGTCCCGGTTTAGGGG
Phenotype
MIM: 103071
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCY2 PubMed Links

Gene Details

Gene
ADCY2
Gene Name
adenylate cyclase 2
There are no transcripts associated with this gene.

Gene
C5orf49
Gene Name
chromosome 5 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001089584.2 828 Missense Mutation GAC,GCC D120A NP_001083053.1
XM_011513963.2 828 Missense Mutation GAC,GCC D88A XP_011512265.1

View Full Product Details