Product Details

SNP ID
rs138627201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36247934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGAATGACCTAACCACTTACC[G/T]CTTCCAAGTTCTGAATTTTACAACA
Phenotype
MIM: 612121
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNPLA1 PubMed Links

Gene Details

Gene
PNPLA1
Gene Name
patatin like phospholipase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145716.2 Intron NP_001139188.1
NM_001145717.1 Intron NP_001139189.2
NM_173676.2 Intron NP_775947.2
XM_011514519.2 Intron XP_011512821.1
XM_011514520.2 Intron XP_011512822.1
XM_017010776.1 Intron XP_016866265.1
XM_017010777.1 Intron XP_016866266.1
XM_017010778.1 Intron XP_016866267.1
XM_017010779.1 Intron XP_016866268.1

View Full Product Details