Product Details

SNP ID
rs138989581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:146614506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTTTCCAGAATTTTATATAAA[C/T]GGAATCAAACCCATGTACACATTTT
Phenotype
MIM: 614630
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGB PubMed Links

Gene Details

Gene
ADGB
Gene Name
androglobin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024694.3 Intron NP_078970.3
XM_006715566.3 Intron XP_006715629.1
XM_011536125.2 Intron XP_011534427.1
XM_011536126.1 Intron XP_011534428.1
XM_011536127.2 Intron XP_011534429.1
XM_011536128.2 Intron XP_011534430.1
XM_011536130.2 Intron XP_011534432.1
XM_011536131.2 Intron XP_011534433.1
XM_011536132.2 Intron XP_011534434.1
XM_011536134.2 Intron XP_011534436.1
XM_011536135.2 Intron XP_011534437.1
XM_017011314.1 Intron XP_016866803.1
XM_017011315.1 Intron XP_016866804.1

View Full Product Details