Product Details

SNP ID
rs139874883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCGGCTGAACCACGAGGGCAAG[A/G]TAGTCCTCAGGATGCGCAGCCTCAA
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 541 Missense Mutation ATA,GTA I65V NP_001025034.1
NM_001317342.1 541 Missense Mutation ATA,GTA I84V NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 541 Intron NP_001189479.1
NM_182552.4 541 Intron NP_872358.4
XM_011535682.2 541 Intron XP_011533984.1
XM_011535684.2 541 Intron XP_011533986.1
XM_011535685.2 541 Intron XP_011533987.1
XM_011535687.2 541 Intron XP_011533989.1
XM_011535688.2 541 Intron XP_011533990.1
XM_011535689.2 541 Intron XP_011533991.1
XM_011535690.2 541 Intron XP_011533992.1
XM_011535691.2 541 Intron XP_011533993.1
XM_011535692.2 541 Intron XP_011533994.1
XM_011535693.2 541 Intron XP_011533995.1
XM_011535694.2 541 Intron XP_011533996.1
XM_011535696.2 541 Intron XP_011533998.1
XM_011535697.2 541 Intron XP_011533999.1
XM_017010658.1 541 Intron XP_016866147.1
XM_017010659.1 541 Intron XP_016866148.1
XM_017010660.1 541 Intron XP_016866149.1
XM_017010661.1 541 Intron XP_016866150.1
XM_017010662.1 541 Intron XP_016866151.1
XM_017010663.1 541 Intron XP_016866152.1
XM_017010664.1 541 Intron XP_016866153.1
XM_017010665.1 541 Intron XP_016866154.1
XM_017010666.1 541 Intron XP_016866155.1
XM_017010667.1 541 Intron XP_016866156.1
XM_017010668.1 541 Intron XP_016866157.1
XM_017010669.1 541 Intron XP_016866158.1
XM_017010670.1 541 Intron XP_016866159.1
XM_017010671.1 541 Intron XP_016866160.1
XM_017010672.1 541 Intron XP_016866161.1
XM_017010673.1 541 Intron XP_016866162.1
XM_017010674.1 541 Intron XP_016866163.1
XM_017010675.1 541 Intron XP_016866164.1

View Full Product Details