Product Details

SNP ID
rs141525894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32195685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCGCAGGGTTGGGCCGAGGCC[C/T]GCGCATGCCTGCAGAAAACCTACGG
Phenotype
MIM: 164951 MIM: 176311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPSM3 PubMed Links

Gene Details

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276501.1 5903 Intron NP_001263430.1
NM_022107.2 5903 Intron NP_071390.1
Gene
NOTCH4
Gene Name
notch 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004557.3 5903 Missense Mutation NP_004548.3
Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

View Full Product Details