Product Details

SNP ID
rs143773461
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151365541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTCAGTCTAGTAAGACGGTTT[C/T]TTGTTCCTTTTTGATGGAAGCTAAC
Phenotype
MIM: 604698 MIM: 616595
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AKAP12 PubMed Links

Gene Details

Gene
AKAP12
Gene Name
A-kinase anchoring protein 12
There are no transcripts associated with this gene.

Gene
ZBTB2
Gene Name
zinc finger and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020861.2 1900 Missense Mutation AAA,GAA K509E NP_065912.1
XM_005267076.2 1900 Missense Mutation AAA,GAA K509E XP_005267133.1
XM_011536004.2 1900 Missense Mutation AAA,GAA K509E XP_011534306.1
XM_017011137.1 1900 Missense Mutation AAA,GAA K430E XP_016866626.1

View Full Product Details