Product Details

SNP ID
rs144358792
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32949615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCAGGGAGAAAGCCTCACCCCA[A/G]TAGGCAATTGCTGTGTAGCGGTCAA
Phenotype
MIM: 142855 MIM: 142856
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DMA PubMed Links

Gene Details

Gene
HLA-DMA
Gene Name
major histocompatibility complex, class II, DM alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006120.3 734 Silent Mutation TAC,TAT Y216Y NP_006111.2
Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
There are no transcripts associated with this gene.

View Full Product Details