Product Details

SNP ID
rs148012848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32195513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCACAGTCAAGTTGAGGTGAT[C/G]CCCGCTCCGGGGACGGAGTAAGGCA
Phenotype
MIM: 164951 MIM: 176311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPSM3 PubMed Links

Gene Details

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276501.1 6075 Intron NP_001263430.1
NM_022107.2 6075 UTR 5 NP_071390.1
Gene
NOTCH4
Gene Name
notch 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004557.3 6075 Missense Mutation GCA,GGA A1979G NP_004548.3
Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

View Full Product Details