Product Details

SNP ID
rs148926387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36400488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTATGATCAATGTTGTCCCCA[A/G]TGTGTCTCAGCTGCATGGCCAACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PXT1 PubMed Links

Gene Details

Gene
PXT1
Gene Name
peroxisomal, testis specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152990.3 750 Missense Mutation ACT,ATT T89I NP_694535.2
XM_011514400.2 750 Missense Mutation ACT,ATT T94I XP_011512702.1

View Full Product Details