Product Details
- SNP ID
-
rs139075508
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149103783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGAAGCCTTTGCCACACTCGGG[A/G]CACTGGAAGGGCCTCTCTCCACTGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF425
PubMed Links
Gene Details
- Gene
- ZNF425
- Gene Name
- zinc finger protein 425
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001661.2 |
2221 |
Silent Mutation |
TGC,TGT |
C696C |
NP_001001661.1 |
View Full Product Details