Product Details

SNP ID
rs139636296
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74232119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCTCTGCTCTAACTGGCCACC[A/G]GGGCCATTGTCTTCTGACACAGCCT
Phenotype
MIM: 605719 MIM: 600404
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LAT2 PubMed Links

Gene Details

Gene
LAT2
Gene Name
linker for activation of T-cells family member 2
There are no transcripts associated with this gene.

Gene
RFC2
Gene Name
replication factor C subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278791.1 873 Intron NP_001265720.1
NM_001278792.1 873 Intron NP_001265721.1
NM_001278793.1 873 Intron NP_001265722.1
NM_002914.4 873 Intron NP_002905.2
NM_181471.2 873 Intron NP_852136.1
XM_006716080.2 873 Missense Mutation CCG,CTG P214L XP_006716143.1
XM_017012492.1 873 Missense Mutation CCG,CTG P250L XP_016867981.1
XM_017012493.1 873 Missense Mutation CCG,CTG P214L XP_016867982.1

View Full Product Details