Product Details

SNP ID
rs142147954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151082246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGAATCATTGAGGAATTTCAG[C/T]CGTAGCACGAGGGGCTCCTGCGGGG
Phenotype
MIM: 616813 MIM: 606965 MIM: 109280 MIM: 614792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGAP3 PubMed Links

Gene Details

Gene
AGAP3
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
There are no transcripts associated with this gene.

Gene
FASTK
Gene Name
Fas activated serine/threonine kinase
There are no transcripts associated with this gene.

Gene
SLC4A2
Gene Name
solute carrier family 4 member 2
There are no transcripts associated with this gene.

Gene
TMUB1
Gene Name
transmembrane and ubiquitin like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136044.1 448 Silent Mutation CGA,CGG R106R NP_001129516.1
NM_031434.3 448 Silent Mutation CGA,CGG R106R NP_113622.1

View Full Product Details