Product Details

SNP ID
rs143198008
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:886134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTCTTCTTCAGCCATGGCCA[A/G]GTAAGCCGTCTTGCTTCCTCCTGCA
Phenotype
MIM: 612056
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GET4 PubMed Links

Gene Details

Gene
GET4
Gene Name
golgi to ER traffic protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015949.2 326 Silent Mutation CAA,CAG Q78Q NP_057033.2

View Full Product Details