Product Details

SNP ID
rs143968240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27525999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTCAATTAAGCTAGTTAGCAGA[A/G]ACTATCAGTGGCTTGCAGAAAAAAA
Phenotype
MIM: 608475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIBADH PubMed Links

Gene Details

Gene
HIBADH
Gene Name
3-hydroxyisobutyrate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152740.3 1469 UTR 3 NP_689953.1
XM_017011719.1 1469 UTR 3 XP_016867208.1

View Full Product Details