Product Details
- SNP ID
-
rs149948419
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73769800 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCGGCCAGCAGGATGGCCACCA[C/G]GATGAGGGCGCGGGCCGCCTGAAGG
- Phenotype
-
MIM: 602910
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN3
PubMed Links
Gene Details
- Gene
- CLDN3
- Gene Name
- claudin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306.3 |
471 |
Missense Mutation |
CTG,GTG |
L84V |
NP_001297.1 |
View Full Product Details