Product Details
- SNP ID
-
rs150412959
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149104078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACTGTGCAGCCTCAGGTGCTCG[A/G]TGAGCTGAGACTGATGCGTGTAGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF425
PubMed Links
Gene Details
- Gene
- ZNF425
- Gene Name
- zinc finger protein 425
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001661.2 |
1926 |
Missense Mutation |
ACC,ATC |
T598I |
NP_001001661.1 |
View Full Product Details