Product Details

SNP ID
rs116475566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:9138648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAGCACAAGACTCTTAGCTTG[A/C]AAAGCCTGACAACTCTGCCTCCAAA
Phenotype
MIM: 610541
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R3B PubMed Links

Gene Details

Gene
PPP1R3B
Gene Name
protein phosphatase 1 regulatory subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201329.1 3122 UTR 3 NP_001188258.1
NM_024607.3 3122 UTR 3 NP_078883.2
XM_006716253.3 3122 UTR 3 XP_006716316.1
XM_017013835.1 3122 UTR 3 XP_016869324.1

View Full Product Details