Product Details

SNP ID
rs138228101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56163341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTAATCATATCTAGTTGACA[C/T]AGAAACAAACAATCTTTTTCTAGGG
Phenotype
MIM: 603026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAG1 PubMed Links

Gene Details

Gene
PLAG1
Gene Name
PLAG1 zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114634.1 4906 UTR 3 NP_001108106.1
NM_001114635.1 4906 UTR 3 NP_001108107.1
NM_002655.2 4906 UTR 3 NP_002646.2
XM_011517544.2 4906 UTR 3 XP_011515846.1
XM_017013576.1 4906 UTR 3 XP_016869065.1
XM_017013577.1 4906 UTR 3 XP_016869066.1

View Full Product Details