Product Details

SNP ID
rs139144351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123325898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGATTTCAATTTACATACTTTTA[A/T]TCGCTCATGATCCACAACAAGTGAG
Phenotype
MIM: 611941
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATAD2 PubMed Links

Gene Details

Gene
ATAD2
Gene Name
ATPase family, AAA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014109.3 2525 Missense Mutation ATA,TTA I1333L NP_054828.2
XM_011516994.2 2525 Missense Mutation ATA,TTA I1203L XP_011515296.1
XM_011516995.2 2525 Intron XP_011515297.1
XM_011516996.2 2525 Missense Mutation ATA,TTA I792L XP_011515298.1

View Full Product Details