Product Details

SNP ID
rs141222335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94130690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCACAGTGGGTATCCCAGTCTG[A/G]TGACCTGCTGGCCGGAAACAACTTC
Phenotype
MIM: 603017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH17 PubMed Links

Gene Details

Gene
CDH17
Gene Name
cadherin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144663.1 2460 Silent Mutation CAC,CAT H778H NP_001138135.1
NM_004063.3 2460 Silent Mutation CAC,CAT H778H NP_004054.3
XM_011516790.1 2460 Silent Mutation CAC,CAT H698H XP_011515092.1

View Full Product Details