Product Details

SNP ID
rs143408057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144504689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGAGGCCCATCACCTTCCTGC[A/G]CCAGGTGAGGCTCCTGCACTGCCCG
Phenotype
MIM: 138200 MIM: 609172 MIM: 603780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPT PubMed Links

Gene Details

Gene
GPT
Gene Name
glutamic--pyruvic transaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005309.2 1161 Missense Mutation CAC,CGC H83R NP_005300.1
XM_011516993.2 1161 Missense Mutation CAC,CGC H83R XP_011515295.1
Gene
LOC101928953
Gene Name
uncharacterized LOC101928953
There are no transcripts associated with this gene.

Gene
MFSD3
Gene Name
major facilitator superfamily domain containing 3
There are no transcripts associated with this gene.

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
There are no transcripts associated with this gene.

Gene
RECQL4
Gene Name
RecQ like helicase 4
There are no transcripts associated with this gene.

View Full Product Details