Product Details

SNP ID
rs147178230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109087893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCATTGAGAGGTACATAGCAAT[A/C]TGTCACCCCATCAAAGCCCAGTTTC
Phenotype
MIM: 188545
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 470 Silent Mutation ATA,ATC I127I NP_003292.1
XM_011517263.2 470 Silent Mutation ATA,ATC I127I XP_011515565.1

View Full Product Details