Product Details

SNP ID
rs149569704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30797483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGGAAAAGGCCGAGGAGATG[A/C]AGAAACACAGGTCATATGAATCTAG
Phenotype
MIM: 176916
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPP2CB PubMed Links

Gene Details

Gene
PPP2CB
Gene Name
protein phosphatase 2 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009552.1 Intron NP_001009552.1

View Full Product Details