Product Details

SNP ID
rs150302473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:36791333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTTGTTAATTACATAACTGAC[G/T]GTGTTTCTCTATTGCTTTGTGGCTA
Phenotype
MIM: 615215
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNU1 PubMed Links

Gene Details

Gene
KCNU1
Gene Name
potassium calcium-activated channel subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031836.2 Intron NP_001027006.2
XM_011544424.1 Intron XP_011542726.1
XM_011544426.1 Intron XP_011542728.1
XM_011544427.1 Intron XP_011542729.1
XM_017013144.1 Intron XP_016868633.1

View Full Product Details