Product Details

SNP ID
rs150785651
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37844737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTAAACCTAAGGAATTATTTCCC[A/C]CCTCCCCTTCTCCTTGCCCCTGTCC
Phenotype
MIM: 606823 MIM: 607013
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ADGRA2 PubMed Links

Gene Details

Gene
ADGRA2
Gene Name
adhesion G protein-coupled receptor A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032777.9 1143 Intron NP_116166.9
XM_005273471.3 1143 Intron XP_005273528.1
XM_011544481.2 1143 Intron XP_011542783.1
XM_011544482.2 1143 Intron XP_011542784.1
XM_011544483.2 1143 Intron XP_011542785.1
Gene
BRF2
Gene Name
BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018310.3 1143 Missense Mutation GGG,GTG G338V NP_060780.2

View Full Product Details