Product Details

SNP ID
rs138321845
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92457277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTATATTAAATGGACATTACCA[A/G]TTACTAATTATTACATTCCAAAGTT
Phenotype
MIM: 608135 MIM: 611505
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ASPN PubMed Links

Gene Details

Gene
ASPN
Gene Name
asporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193335.1 1218 UTR 3 NP_001180264.1
NM_017680.4 1218 UTR 3 NP_060150.4
Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 1218 Intron NP_001012267.1
NM_001286969.1 1218 Intron NP_001273898.1
NM_001286971.1 1218 Intron NP_001273900.1
XM_011518685.2 1218 Intron XP_011516987.1
XM_011518689.1 1218 Intron XP_011516991.1
XM_017014715.1 1218 Intron XP_016870204.1

View Full Product Details