Product Details

SNP ID
rs138366871
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37512619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCGGGCTGTGATCCTCGTTGC[C/T]ATGGCTGTCACCTTCTGCCCATTGT
Phenotype
MIM: 609092
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXO10 PubMed Links

Gene Details

Gene
FBXO10
Gene Name
F-box protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012166.2 3165 Missense Mutation ATA,ATG I933M NP_036298.2
XM_005251439.4 3165 Intron XP_005251496.1
XM_006716754.3 3165 Missense Mutation ATA,ATG I458M XP_006716817.1
XM_017014618.1 3165 Missense Mutation ATA,ATG I991M XP_016870107.1
XM_017014619.1 3165 Missense Mutation ATA,ATG I933M XP_016870108.1
Gene
POLR1E
Gene Name
RNA polymerase I subunit E
There are no transcripts associated with this gene.

View Full Product Details