Product Details

SNP ID
rs141834891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:12694063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTGCTACTTTTTCAGCAGGCC[C/T]GGGCTCAATTCCCAAGACAGTGTGC
Phenotype
MIM: 115501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LURAP1L-AS1 PubMed Links

Gene Details

Gene
LURAP1L-AS1
Gene Name
LURAP1L antisense RNA 1
There are no transcripts associated with this gene.

Gene
TYRP1
Gene Name
tyrosinase related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000550.2 245 Missense Mutation CGG,TGG R23W NP_000541.1

View Full Product Details