Product Details

SNP ID
rs142346579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133473505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCGCCAAGTCCCCCCAGGACT[C/T]GCTTTCCAGGCCCGCAGTGCTGTTG
Phenotype
MIM: 613104 MIM: 606813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACFD1 PubMed Links

Gene Details

Gene
CACFD1
Gene Name
calcium channel flower domain containing 1
There are no transcripts associated with this gene.

Gene
SLC2A6
Gene Name
solute carrier family 2 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145099.1 1199 Intron NP_001138571.1
NM_017585.3 1199 Missense Mutation AAG,GAG K378E NP_060055.2
XM_011518189.2 1199 Missense Mutation AAG,GAG K172E XP_011516491.2
XM_017014236.1 1199 Intron XP_016869725.1
XM_017014237.1 1199 Missense Mutation AAG,GAG K134E XP_016869726.1
XM_017014238.1 1199 Missense Mutation AAG,GAG K134E XP_016869727.1

View Full Product Details