Product Details

SNP ID
rs142501660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137607425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCGGTGACAGTCAGCACCCAG[A/C]CGTGAGGACACCGGCTGTCTAAAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARRDC1 PubMed Links

Gene Details

Gene
ARRDC1
Gene Name
arrestin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317968.1 Intron NP_001304897.1
NM_152285.3 Intron NP_689498.1
XM_005266119.1 Intron XP_005266176.1
XM_006717320.3 Intron XP_006717383.1
XM_006717321.3 Intron XP_006717384.1
XM_006717322.3 Intron XP_006717385.1
XM_017015288.1 Intron XP_016870777.1
Gene
ARRDC1-AS1
Gene Name
ARRDC1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details