Product Details

SNP ID
rs142547455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131290125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGCAGCTCGGGCAGAAAGGCCT[C/T]GGGCCCATCAGCACAGCCCCCACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM78A PubMed Links

Gene Details

Gene
FAM78A
Gene Name
family with sequence similarity 78 member A
There are no transcripts associated with this gene.

Gene
PLPP7
Gene Name
phospholipid phosphatase 7 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032728.3 444 Missense Mutation TCG,TTG S43L NP_116117.3
XM_005272230.3 444 Missense Mutation TCG,TTG S43L XP_005272287.1
XM_005272231.1 444 Missense Mutation TCG,TTG S43L XP_005272288.1

View Full Product Details