Product Details

SNP ID
rs142863012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132945627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGGCAGAGTGAGATGAAGTC[A/G]AACTGTGGGATATGAAAGGCATTTT
Phenotype
MIM: 604383 MIM: 605284
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFI1B PubMed Links

Gene Details

Gene
GFI1B
Gene Name
growth factor independent 1B transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135031.1 83 Intron NP_001128503.1
NM_004188.6 83 UTR 5 NP_004179.3
XM_006717297.3 83 Intron XP_006717360.1
XM_011519068.2 83 Intron XP_011517370.1
XM_011519069.2 83 Intron XP_011517371.1
XM_011519070.2 83 Intron XP_011517372.1
XM_017015175.1 83 Intron XP_016870664.1
XM_017015176.1 83 Intron XP_016870665.1
Gene
MIR548AW
Gene Name
microRNA 548aw
There are no transcripts associated with this gene.

Gene
TSC1
Gene Name
tuberous sclerosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000368.4 83 Intron NP_000359.1
NM_001162426.1 83 Intron NP_001155898.1
NM_001162427.1 83 Intron NP_001155899.1
XM_005272211.1 83 Intron XP_005272268.1
XM_006717271.1 83 Intron XP_006717334.1
XM_011518979.2 83 Intron XP_011517281.1
XM_017015096.1 83 Intron XP_016870585.1
XM_017015097.1 83 Intron XP_016870586.1
XM_017015098.1 83 Intron XP_016870587.1
XM_017015099.1 83 Intron XP_016870588.1
XM_017015100.1 83 Intron XP_016870589.1
XM_017015101.1 83 Intron XP_016870590.1

View Full Product Details