Product Details

SNP ID
rs146553903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37440252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTTTAAATGTGCCACTTTTA[A/G]TATCAATTACAAACTACTTTCTGAC
Phenotype
MIM: 604296 MIM: 616590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRHPR PubMed Links

Gene Details

Gene
GRHPR
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
There are no transcripts associated with this gene.

Gene
ZBTB5
Gene Name
zinc finger and BTB domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014872.2 2500 UTR 3 NP_055687.1
XM_005251634.1 2500 UTR 3 XP_005251691.1

View Full Product Details