Product Details

SNP ID
rs146882840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34254386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCACAGGGTCTGGCTCTAAGA[C/T]GGCACTGTTCCCTCAGGGGCTGCGG
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 2825 Silent Mutation CCA,CCG P1367P NP_919289.2
XM_005251451.2 2825 Silent Mutation CCA,CCG P1006P XP_005251508.1
XM_006716764.2 2825 Silent Mutation CCA,CCG P909P XP_006716827.1
XM_011517861.1 2825 Silent Mutation CCA,CCG P1398P XP_011516163.1
XM_011517863.2 2825 Silent Mutation CCA,CCG P1367P XP_011516165.1
XM_017014697.1 2825 Silent Mutation CCA,CCG P1367P XP_016870186.1
XM_017014698.1 2825 Silent Mutation CCA,CCG P1078P XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details