Product Details

SNP ID
rs147580228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110244211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTCCTTATTGGCTCCAGAAAA[C/T]TCACCCACCTGTTAAGAGAATATTG
Phenotype
MIM: 187700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXN PubMed Links

Gene Details

Gene
TXN
Gene Name
thioredoxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244938.1 469 Silent Mutation GAA,GAG E68E NP_001231867.1
NM_003329.3 469 Silent Mutation GAA,GAG E88E NP_003320.2

View Full Product Details