Product Details

SNP ID
rs147996634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21802646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCGCCCCTGGTCTCCGCACTG[C/G]TCACTCCCGCGCAGTGAGGTTGGCA
Phenotype
MIM: 156540
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTAP PubMed Links

Gene Details

Gene
MTAP
Gene Name
methylthioadenosine phosphorylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002451.3 11 UTR 5 NP_002442.2

View Full Product Details