Product Details

SNP ID
rs148332697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99217966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTTTGGTAACATATCGGTAGA[A/G]CTGTTCTGTAAATGCTTCAGGGGAA
Phenotype
MIM: 607905 MIM: 609214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG2 PubMed Links

Gene Details

Gene
ALG2
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033087.3 1289 Missense Mutation CTC,TTC L407F NP_149078.1
Gene
SEC61B
Gene Name
Sec61 translocon beta subunit
There are no transcripts associated with this gene.

View Full Product Details