Product Details

SNP ID
rs148732465
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127621544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCAAACTCATATACAAATGTC[A/G]CAGCAGCATTATTCATAACAGCCAA
Phenotype
MIM: 602926
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
STXBP1 PubMed Links

Gene Details

Gene
STXBP1
Gene Name
syntaxin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032221.3 Intron NP_001027392.1
NM_003165.3 Intron NP_003156.1

View Full Product Details