Product Details

SNP ID
rs151088980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136993215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGACACTGGGCCTGGCAAAACG[C/T]GTGTGGAGCCTGGAGCGGCGACTGG
Phenotype
MIM: 616315 MIM: 606533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
There are no transcripts associated with this gene.

Gene
LCNL1
Gene Name
lipocalin like 1
There are no transcripts associated with this gene.

View Full Product Details