Product Details

SNP ID
rs111653090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153909471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCAGCTCCGATGCCAGGAGGC[C/T]CTCGGGACTGCTCCCAGACTCCCCT
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164741.1 2657 Missense Mutation AGC,GGC S867G NP_001158213.1
NM_001666.4 2657 Missense Mutation AGC,GGC S827G NP_001657.3
Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
There are no transcripts associated with this gene.

View Full Product Details