Product Details

SNP ID
rs139621376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:22273452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCGCTTCGCTTGAAAAAGTTC[A/G]TCCTCATATTGCTCCGCCACAAACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTCHD1-AS PubMed Links

Gene Details

Gene
PTCHD1-AS
Gene Name
PTCHD1 antisense RNA (head to head)
There are no transcripts associated with this gene.

Gene
ZNF645
Gene Name
zinc finger protein 645
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152577.3 540 Missense Mutation CAT,CGT H154R NP_689790.1

View Full Product Details