Product Details

SNP ID
rs140222626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49256847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAGGAGCTCTGGGGCACAGCC[A/G]AAAGGGTGCTGGGGGGACAGAGGGT
Phenotype
MIM: 300859 MIM: 300292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC22 PubMed Links

Gene Details

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
There are no transcripts associated with this gene.

Gene
FOXP3
Gene Name
forkhead box P3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114377.1 739 Missense Mutation TCG,TTG S149L NP_001107849.1
NM_014009.3 739 Missense Mutation TCG,TTG S184L NP_054728.2
XM_006724533.2 739 Missense Mutation TCG,TTG S207L XP_006724596.2
XM_011543916.2 739 Missense Mutation TCG,TTG S257L XP_011542218.1
XM_017029565.1 739 Missense Mutation TCG,TTG S269L XP_016885054.1
XM_017029566.1 739 Missense Mutation TCG,TTG S257L XP_016885055.1
XM_017029567.1 739 Missense Mutation TCG,TTG S166L XP_016885056.1

View Full Product Details