Product Details

SNP ID
hCV175346818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49251439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGCCCCCTTCTCGCTCTCCAC[C/T]CGCACAAAGCACTTGTGCAGACTCA
Phenotype
MIM: 300859 MIM: 300292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC22 PubMed Links

Gene Details

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
There are no transcripts associated with this gene.

Gene
FOXP3
Gene Name
forkhead box P3
There are no transcripts associated with this gene.

View Full Product Details