Product Details

SNP ID
rs143619652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153909533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCGCCCACCACCTGCTTCTCC[A/G]TCCTGCGGTGGGAAGGACCGGCCTG
Phenotype
MIM: 300023 MIM: 300538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP4 PubMed Links

Gene Details

Gene
ARHGAP4
Gene Name
Rho GTPase activating protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164741.1 2595 Missense Mutation ACG,ATG T846M NP_001158213.1
NM_001666.4 2595 Missense Mutation ACG,ATG T806M NP_001657.3
Gene
AVPR2
Gene Name
arginine vasopressin receptor 2
There are no transcripts associated with this gene.

View Full Product Details