Product Details

SNP ID
rs146302543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47977292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCTAAAAGCTTTTCCACATTC[A/G]GTACATCCAAAAGGTTTCTCTCCCG
Phenotype
MIM: 314993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF182 PubMed Links

Gene Details

Gene
ZNF182
Gene Name
zinc finger protein 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007088.1 1047 Silent Mutation ACC,ACT T246T NP_001007089.1
NM_001178099.1 1047 Silent Mutation ACC,ACT T265T NP_001171570.1
NM_006962.1 1047 Silent Mutation ACC,ACT T265T NP_008893.1

View Full Product Details