Product Details

SNP ID
rs146438992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:139726822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTCACAAATAGGGTGATTACA[C/T]TGAGTGCTAGGAATACTGCATGGTC
Phenotype
MIM: 300516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP11C PubMed Links

Gene Details

Gene
ATP11C
Gene Name
ATPase phospholipid transporting 11C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010986.2 6259 UTR 3 NP_001010986.1
NM_173694.4 6259 UTR 3 NP_775965.2
XM_017029439.1 6259 UTR 3 XP_016884928.1
XM_017029440.1 6259 UTR 3 XP_016884929.1
XM_017029441.1 6259 UTR 3 XP_016884930.1
XM_017029442.1 6259 UTR 3 XP_016884931.1
XM_017029443.1 6259 UTR 3 XP_016884932.1
XM_017029444.1 6259 UTR 3 XP_016884933.1
XM_017029445.1 6259 Intron XP_016884934.1
XM_017029446.1 6259 UTR 3 XP_016884935.1
XM_017029447.1 6259 UTR 3 XP_016884936.1
XM_017029448.1 6259 UTR 3 XP_016884937.1
XM_017029449.1 6259 UTR 3 XP_016884938.1

View Full Product Details