Product Details
- SNP ID
-
rs147304585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:103063132 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTAACGCGGAACCGCCTACGATTT[C/T]CTCTAGGCACACAGTATTCACCAGC
- Phenotype
-
MIM: 300690
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BEX1
PubMed Links
Gene Details
- Gene
- BEX1
- Gene Name
- brain expressed X-linked 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018476.3 |
383 |
Missense Mutation |
GAA,GGA |
E48G |
NP_060946.3 |
View Full Product Details