Product Details

SNP ID
rs148236809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130209689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGCTTCTTGAAAATACAAAAC[C/T]GTTTACCTGGATGGTTGCTATGAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF280C PubMed Links

Gene Details

Gene
ZNF280C
Gene Name
zinc finger protein 280C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017666.4 2209 Missense Mutation CAG,CGG Q669R NP_060136.1
XM_006724765.3 2209 Missense Mutation CAG,CGG Q620R XP_006724828.1

View Full Product Details