Product Details

SNP ID
rs148814895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47572070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGCACGATGAACGAGGACCAC[A/G]TATGCCAAGCACGGCACAAAAGGAG
Phenotype
MIM: 311010 MIM: 313440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARAF PubMed Links

Gene Details

Gene
ARAF
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256196.1 3041 Intron NP_001243125.1
NM_001256197.1 3041 Intron NP_001243126.1
NM_001654.4 3041 Intron NP_001645.1
XM_006724529.2 3041 Intron XP_006724592.1
XM_011543906.2 3041 Intron XP_011542208.1
XM_011543908.2 3041 Intron XP_011542210.1
XM_011543909.2 3041 Intron XP_011542211.1
Gene
SYN1
Gene Name
synapsin I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006950.3 3041 UTR 3 NP_008881.2
NM_133499.2 3041 UTR 3 NP_598006.1

View Full Product Details