Product Details

SNP ID
rs149205899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:3066626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGGCTGAATGTAATGTTTTGT[C/T]GTTGATGTGCTGTTGTTTTTCTTTT
Phenotype
MIM: 300003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSF PubMed Links

Gene Details

Gene
ARSF
Gene Name
arylsulfatase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201538.1 Intron NP_001188467.1
NM_001201539.1 Intron NP_001188468.1
NM_004042.4 Intron NP_004033.2
XM_011545522.1 Intron XP_011543824.1
XM_011545523.1 Intron XP_011543825.1
XM_011545524.1 Intron XP_011543826.1
XM_017029527.1 Intron XP_016885016.1
XM_017029528.1 Intron XP_016885017.1

View Full Product Details